Tisk…

2,4-Dibromo-5-methoxybenzaldehyde 99%

Dodavatel: Apollo Scientific
Varování

OR55242-1G OR55242-5G
APOSOR55242-1GEA 7210 CZK
APOSOR55242-1G APOSOR55242-5G
2,4-Dibromo-5-methoxybenzaldehyde 99%
2,4-Dibromo-5-methoxybenzaldehyde
Vzorec: C₈H₆Br₂O₂
MW: 293,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27935321
CAS číslo: 99615-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR