2,4-Dibromo-5-hydroxybenzaldehyde 99%
Dodavatel: Apollo Scientific
|
Varování
|
OR55241-1G
OR55241-5G
OR55241-25G
APOSOR55241-1GEA
6620
CZK
APOSOR55241-1G
APOSOR55241-5G
APOSOR55241-25G
2,4-Dibromo-5-hydroxybenzaldehyde 99%
2,4-Dibromo-5-hydroxybenzaldehyde
Vzorec:
C₇H₄Br₂O₂ MW: 279,913 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08673515 CAS číslo: 3111-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...