Tisk…

3-(1H-Pyrazol-3-yl)benzonitrile

Dodavatel: Apollo Scientific

OR55221-1G OR55221-250MG
APOSOR55221-1GEA 7440 CZK
APOSOR55221-1G APOSOR55221-250MG
3-(1H-Pyrazol-3-yl)benzonitrile
3-(1H-Pyrazol-3-yl)benzonitrile
Vzorec: C₁₀H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD09860597
CAS číslo: 149739-51-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR