Ethyl 2-(methylsulphonyl)pyrimidine-5-carboxylate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR55185-100MG
OR55185-1G
OR55185-250MG
APOSOR55185-100MGEA
1740
CZK
APOSOR55185-100MG
APOSOR55185-1G
APOSOR55185-250MG
Ethyl 2-(methylsulphonyl)pyrimidine-5-carboxylate 95%
Ethyl 2-(methylsulphonyl)pyrimidine-5-carboxylate
Vzorec:
C₈H₁₀N₂O₄S Storage Temperature: Ambient |
MDL Number:
MFCD07366751 CAS číslo: 148550-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...