Tisk…

Methyl CBZ-amino(diethoxyphosphoryl)acetate

Dodavatel: Apollo Scientific
Varování

OR55163-1G OR55163-5G OR55163-25G
APOSOR55163-1GEA 992 CZK
APOSOR55163-1G APOSOR55163-5G APOSOR55163-25G
Methyl CBZ-amino(diethoxyphosphoryl)acetate
Methyl CBZ-amino(diethoxyphosphoryl)acetate
Vzorec: C₁₅H₂₂NO₇P
Storage Temperature: Refrigerator
MDL Number: MFCD09878835
CAS číslo: 114684-69-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR