Tisk…

3-(4-Bromophenyl)-5-methylisoxazole-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR55102-100MG OR55102-250MG
APOSOR55102-100MGEA 1740 CZK
APOSOR55102-100MG APOSOR55102-250MG
3-(4-Bromophenyl)-5-methylisoxazole-4-carboxylic acid
3-(4-Bromophenyl)-5-methylisoxazole-4-carboxylic acid
Vzorec: C₁₁H₈BrNO₃
MW: 282,09 g/mol
Bod tání: 216…218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07376078
CAS číslo: 91182-58-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR