Tisk…

Methyl 2-(4-(hydroxymethyl)phenyl)acetate 95%

Dodavatel: Apollo Scientific
Varování

OR55070-1G OR55070-250MG OR55070-5G
APOSOR55070-1GEA 4770 CZK
APOSOR55070-1G APOSOR55070-250MG APOSOR55070-5G
Methyl 2-(4-(hydroxymethyl)phenyl)acetate 95%
Methyl 2-(4-(hydroxymethyl)phenyl)acetate
Vzorec: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061910
CAS číslo: 155380-11-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR