Tisk…

Potassium oxirane-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR54835-1G OR54835-250MG
APOSOR54835-1GEA 11380 CZK
APOSOR54835-1G APOSOR54835-250MG
Potassium oxirane-2-carboxylate
Potassium oxirane-2-carboxylate
Vzorec: C₃H₃O₃·K
Storage Temperature: Refrigerator
MDL Number: MFCD16293376
CAS číslo: 51877-54-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR