3-Amino-6-bromo-chromen-2-one
Dodavatel: Apollo Scientific
|
Varování
|
OR54833-1G
OR54833-250MG
APOSOR54833-1GEA
17360
CZK
APOSOR54833-1G
APOSOR54833-250MG
3-Amino-6-bromo-chromen-2-one
3-Amino-6-bromo-chromen-2-one
Vzorec:
C₉H₆BrNO₂ MW: 240.053 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29081504 CAS číslo: 33259-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...