Tisk…

4-Cyclopropylthiophene-2-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR54824-1G OR54824-250MG OR54824-5G
APOSOR54824-1GEA 3310 CZK
APOSOR54824-1G APOSOR54824-250MG APOSOR54824-5G
4-Cyclopropylthiophene-2-sulphonyl chloride
4-Cyclopropylthiophene-2-sulphonyl chloride
Vzorec: C₇H₇ClO₂S₂
Storage Temperature: Ambient
MDL Number: MFCD32203986
CAS číslo: 2407349-23-7
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR