2-(Methoxymethyl)aniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR54820-1G
OR54820-250MG
APOSOR54820-1GEA
15510
CZK
APOSOR54820-1G
APOSOR54820-250MG
2-(Methoxymethyl)aniline 95%
2-(Methoxymethyl)aniline
Vzorec:
C₈H₁₁NO MW: 137,179 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09811778 CAS číslo: 62723-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...