Tisk…

2-Bromo-6-methoxybenzeneboronic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-6-methoxyphenylboronic acid

OR54802-1G OR54802-250MG OR54802-5G
APOSOR54802-1GEA 13460 CZK
APOSOR54802-1G APOSOR54802-250MG APOSOR54802-5G
2-Bromo-6-methoxybenzeneboronic acid 95%
2-Bromo-6-methoxybenzeneboronic acid
Vzorec: C₇H₈BBrO₃
MW: 230,852 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23380484
CAS číslo: 352525-79-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR