Tisk…

5-Bromo-3-chloro-2-hydroxybenzaldehyde

Dodavatel: Apollo Scientific
Varování

OR54699-1G OR54699-250MG
APOSOR54699-1GEA 7380 CZK
APOSOR54699-1G APOSOR54699-250MG
5-Bromo-3-chloro-2-hydroxybenzaldehyde
5-Bromo-3-chloro-2-hydroxybenzaldehyde
Vzorec: C₇H₄BrClO₂
MW: 235,462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00807894
CAS číslo: 19652-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR