Tisk…

3,4-Dimethoxy-2-hydroxyphenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

OR54698-1G OR54698-250MG OR54698-5G
APOSOR54698-1GEA 705 CZK
APOSOR54698-1G APOSOR54698-250MG APOSOR54698-5G
3,4-Dimethoxy-2-hydroxyphenacyl bromide
3,4-Dimethoxy-2-hydroxyphenacyl bromide
Vzorec: C₁₀H₁₁BrO₄
MW: 275.096 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32177107
CAS číslo: 18064-92-1
UN: 3262
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR