Tisk…

2-Bromo-3-chlorobenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR54687-100MG OR54687-1G OR54687-250MG
APOSOR54687-100MGEA 7940 CZK
APOSOR54687-100MG APOSOR54687-1G APOSOR54687-250MG
2-Bromo-3-chlorobenzenesulphonyl chloride
2-Bromo-3-chlorobenzenesulphonyl chloride
Vzorec: C₆H₃BrCl₂O₂S
MW: 289,962 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390309
CAS číslo: 1261646-81-4
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR