Indolin-6-ylboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR54681-1G
OR54681-250MG
APOSOR54681-1GEA
29560
CZK
APOSOR54681-1G
APOSOR54681-250MG
Indolin-6-ylboronic acid 95%
Indolin-6-ylboronic acid
Vzorec:
C₈H₁₀BNO₂ MW: 162.982 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18383472 CAS číslo: 1253912-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...