2-Chloro-5-nitronicotinonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR54678-1G
OR54678-5G
OR54678-25G
APOSOR54678-1GEA
1080
CZK
APOSOR54678-1G
APOSOR54678-5G
APOSOR54678-25G
2-Chloro-5-nitronicotinonitrile 97%
2-Chloro-5-nitronicotinonitrile
Vzorec:
C₆H₂ClN₃O₂ Bod tání: 121…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD07345667 CAS číslo: 31309-08-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...