Tisk…

7-Amino-3,4-dihydro-2H-isoquinolin-1-one 97%

Dodavatel: Apollo Scientific
Varování

OR54677-1G OR54677-250MG OR54677-5G
APOSOR54677-1GEA 6390 CZK
APOSOR54677-1G APOSOR54677-250MG APOSOR54677-5G
7-Amino-3,4-dihydro-2H-isoquinolin-1-one 97%
7-Amino-3,4-dihydro-2H-isoquinolin-1-one
Vzorec: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD08437645
CAS číslo: 66491-03-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR