8-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR54671-1G
OR54671-250MG
APOSOR54671-1GEA
32730
CZK
APOSOR54671-1G
APOSOR54671-250MG
8-Bromo-2-oxo-2H-chromene-3-carboxylic acid
8-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Vzorec:
C₁₀H₅BrO₄ Storage Temperature: Ambient |
MDL Number:
MFCD13187196 CAS číslo: 94032-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...