2-Hydroxy-5-cyclopropylbenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR54660-1G
OR54660-250MG
APOSOR54660-1GEA
16530
CZK
APOSOR54660-1G
APOSOR54660-250MG
2-Hydroxy-5-cyclopropylbenzaldehyde
2-Hydroxy-5-cyclopropylbenzaldehyde
Vzorec:
C₁₀H₁₀O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD06802409 CAS číslo: 893738-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...