2-Amino-6-bromo-4-nitrophenol, tech
Dodavatel: Apollo Scientific
|
Varování
|
OR54659-100MG
OR54659-1G
OR54659-250MG
APOSOR54659-100MGEA
3640
CZK
APOSOR54659-100MG
APOSOR54659-1G
APOSOR54659-250MG
2-Amino-6-bromo-4-nitrophenol, tech
2-Amino-6-bromo-4-nitrophenol
Vzorec:
C₆H₅BrN₂O₃ MW: 233,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27975903 CAS číslo: 185335-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...