Tisk…

3-(Cyclopropyl)benzyl alcohol

Dodavatel: Apollo Scientific
Varování

OR54650-1G OR54650-250MG
APOSOR54650-1GEA 2780 CZK
APOSOR54650-1G APOSOR54650-250MG
3-(Cyclopropyl)benzyl alcohol
3-(Cyclopropyl)benzyl alcohol
Vzorec: C₁₀H₁₂O
MW: 148,202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802678
CAS číslo: 893738-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR