3-(Cyclopropyl)benzyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR54648-1G
OR54648-250MG
APOSOR54648-1GEA
14880
CZK
APOSOR54648-1G
APOSOR54648-250MG
3-(Cyclopropyl)benzyl bromide
3-(Cyclopropyl)benzyl bromide
Vzorec:
C₁₀H₁₁Br MW: 211,098 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD15527185 CAS číslo: 1260850-05-2 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...