4-(Cyclopropyl)benzyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-4-cyclopropylbenzene
OR54647-1G
OR54647-250MG
OR54647-5G
APOSOR54647-1GEA
10350
CZK
APOSOR54647-1G
APOSOR54647-250MG
APOSOR54647-5G
4-(Cyclopropyl)benzyl bromide
4-(Cyclopropyl)benzyl bromide
Vzorec:
C₁₀H₁₁Br MW: 211,098 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12024391 CAS číslo: 1150617-57-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...