Methyl 5-(bromomethyl)-2-methylbenzoate 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR54641-1G
OR54641-250MG
OR54641-5G
APOSOR54641-1GEA
17160
CZK
APOSOR54641-1G
APOSOR54641-250MG
APOSOR54641-5G
Methyl 5-(bromomethyl)-2-methylbenzoate 95%
Methyl 5-(bromomethyl)-2-methylbenzoate
Vzorec:
C₁₀H₁₁BrO₂ MW: 243,097 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28148164 CAS číslo: 501362-17-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...