2-Chloro-3-nitroaniline 95+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR54620-100MG
OR54620-250MG
OR54620-1G
APOSOR54620-100MGEA
3060
CZK
APOSOR54620-100MG
APOSOR54620-250MG
APOSOR54620-1G
2-Chloro-3-nitroaniline 95+%
2-Chloro-3-nitroaniline
Vzorec:
C₆H₅ClN₂O₂ MW: 172,569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00800473 CAS číslo: 3970-41-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...