4-Bromo-1-(triisopropylsilyl)-7-azaindole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Bromopyrrolo[2,3-b]pyridin-1-yl)-tri(propan-2-yl)silane
OR54594-1G
OR54594-500MG
APOSOR54594-1GEA
5860
CZK
APOSOR54594-1G
APOSOR54594-500MG
4-Bromo-1-(triisopropylsilyl)-7-azaindole
4-Bromo-1-(triisopropylsilyl)-7-azaindole
Vzorec:
C₁₆H₂₅BrN₂Si MW: 353,373 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11616420 CAS číslo: 640735-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...