Tisk…

Methyl 4-bromo-2-cyanobenzoate

Dodavatel: Apollo Scientific
Varování

OR54532-1G OR54532-250MG OR54532-5G
APOSOR54532-1GEA 4570 CZK
APOSOR54532-1G APOSOR54532-250MG APOSOR54532-5G
Methyl 4-bromo-2-cyanobenzoate
Methyl 4-bromo-2-cyanobenzoate
Vzorec: C₉H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD18398741
CAS číslo: 1223434-15-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR