Tisk…

5-Amino-4-chloro-6-methoxy-2-methylpyrimidine

Dodavatel: Apollo Scientific
Varování

OR54529-100MG OR54529-1G OR54529-250MG OR54529-5G
APOSOR54529-100MGEA 996 CZK
APOSOR54529-100MG APOSOR54529-1G APOSOR54529-250MG APOSOR54529-5G
5-Amino-4-chloro-6-methoxy-2-methylpyrimidine
5-Amino-4-chloro-6-methoxy-2-methylpyrimidine
Vzorec: C₆H₈ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD01837887
CAS číslo: 88474-31-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR