Tisk…

3-Benzoylphenylacetic acid

Dodavatel: Apollo Scientific
Varování

OR54519-1G OR54519-250MG
APOSOR54519-1GEA 8930 CZK
APOSOR54519-1G APOSOR54519-250MG
3-Benzoylphenylacetic acid
3-Benzoylphenylacetic acid
Vzorec: C₁₅H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00046549
CAS číslo: 22071-22-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR