4-Aminobicyclo[2.2.2]octane-1-carboxylic acid hydrochloride
Dodavatel: Apollo Scientific
OR54488-100MG
OR54488-1G
OR54488-250MG
APOSOR54488-100MGEA
7480
CZK
APOSOR54488-100MG
APOSOR54488-1G
APOSOR54488-250MG
4-Aminobicyclo[2.2.2]octane-1-carboxylic acid hydrochloride
4-Aminobicyclo[2.2.2]octane-1-carboxylic acid hydrochloride
Vzorec:
C₉H₁₅NO₂·ClH MW: 205,682 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23701423 CAS číslo: 854214-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...