Tisk…

2-Chloro-3-methylbenzaldehyde 95+%

Dodavatel: Apollo Scientific
Varování

OR54473-250MG OR54473-1G
APOSOR54473-250MGEA 1330 CZK
APOSOR54473-250MG APOSOR54473-1G
2-Chloro-3-methylbenzaldehyde 95+%
2-Chloro-3-methylbenzaldehyde
Vzorec: C₈H₇ClO
Storage Temperature: Refrigerator
MDL Number: MFCD11845907
CAS číslo: 61563-28-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR