4-Cyano-4-{[(ethylsulphanyl)carbonothioyl]sulphanyl}pentanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Cyano-4-(((ethylthio)carbonothioyl)thio)pentanoic acid
OR54471-1G
OR54471-250MG
APOSOR54471-1GEA
39350
CZK
APOSOR54471-1G
APOSOR54471-250MG
4-Cyano-4-{[(ethylsulphanyl)carbonothioyl]sulphanyl}pentanoic acid
4-Cyano-4-{[(ethylsulphanyl)carbonothioyl]sulphanyl}pentanoic acid
Vzorec:
C₉H₁₃NO₂S₃ MW: 263,4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27978407 CAS číslo: 1137725-46-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...