2-Nitro-1H-pyrrole
Dodavatel: Apollo Scientific
|
Varování
|
OR54468-1G
OR54468-250MG
APOSOR54468-1GEA
4140
CZK
APOSOR54468-1G
APOSOR54468-250MG
2-Nitro-1H-pyrrole
2-Nitro-1H-pyrrole
Vzorec:
C₄H₄N₂O₂ MW: 112,087 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12923841 CAS číslo: 5919-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...