Tisk…

(E)-2-Phenylethenesulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: trans-β-Styrenesulphonyl chloride, (E)-2-Phenylethene-1-sulphonyl chloride

OR54457-5G OR54457-10G OR54457-1G
APOSOR54457-5GEA 3040 CZK
APOSOR54457-5G APOSOR54457-10G APOSOR54457-1G
(E)-2-Phenylethenesulphonyl chloride 95%
(E)-2-Phenylethenesulphonyl chloride
Vzorec: C₈H₇ClO₂S
Bod tání: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007456
CAS číslo: 52147-97-4
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR