Tisk…

8-Amino-5-chloroquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloroquinolin-8-amine

OR54420-1G OR54420-250MG
APOSOR54420-1GEA 24770 CZK
APOSOR54420-1G APOSOR54420-250MG
8-Amino-5-chloroquinoline
8-Amino-5-chloroquinoline
Vzorec: C₉H₇ClN₂
MW: 178,618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00461243
CAS číslo: 5432-09-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR