Tisk…

[4-(tert-Butoxycarbonyl)piperazin-1-yl]acetic acid dihydrate

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-(tert-Butoxycarbonyl)piperazin-1-yl)acetic acid dihydrate, 2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]ethanoic acid dihydrate, 2-(1-Boc-piperazin-4-yl)-acetic acid 2H2O, tert-Butyl 4-(carboxymethyl)piperazine-1-carboxylate dihydrate

OR5405-1G OR5405-5G
APOSOR5405-1GEA 1870 CZK
APOSOR5405-1G APOSOR5405-5G
[4-(tert-Butoxycarbonyl)piperazin-1-yl]acetic acid dihydrate
[4-(tert-Butoxycarbonyl)piperazin-1-yl]acetic acid dihydrate
Vzorec: C₁₁H₂₀N₂O₄·2H₂O
MW: 280,318 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03410262
CAS číslo: 1049785-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR