Tisk…

5-Bromo-3-methylbenzo[b]thiophene-2-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Bromo-2-(chlorosulphonyl)-3-methylbenzo[b]thiophene, 5-Bromo-3-methyl-1-benzothiophene-2-sulphonyl chloride

OR5378-1G OR5378-250MG
APOSOR5378-1GEA 10750 CZK
APOSOR5378-1G APOSOR5378-250MG
5-Bromo-3-methylbenzo[b]thiophene-2-sulphonyl chloride
5-Bromo-3-methylbenzo[b]thiophene-2-sulphonyl chloride
Vzorec: C₉H₆BrClO₂S₂
MW: 325,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094671
CAS číslo: 338797-11-8
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR