Tisk…

3-Cyano-2-methylbenzoic acid 95%

Dodavatel: Apollo Scientific

OR53220-1G OR53220-250MG
APOSOR53220-1GEA 32240 CZK
APOSOR53220-1G APOSOR53220-250MG
3-Cyano-2-methylbenzoic acid 95%
3-Cyano-2-methylbenzoic acid
Vzorec: C₉H₇NO₂
MW: 161,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394327
CAS číslo: 3843-99-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR