Tisk…

(S)-Trityl-2-amino-3-hydroxypropionic acid triethyl ammonium salt

Dodavatel: Apollo Scientific
Varování

Synonyms: N-trityl-L-serine triethyl ammonium salt

OR53186-1G OR53186-25G OR53186-5G
APOSOR53186-1GEA 1540 CZK
APOSOR53186-1G APOSOR53186-25G APOSOR53186-5G
(S)-Trityl-2-amino-3-hydroxypropionic acid triethyl ammonium salt
(S)-Trityl-2-amino-3-hydroxypropionic acid triethyl ammonium salt
Vzorec: C₂₂H₂₁NO₃·(C₂H₅)₃N
MW: 448.597 g/mol
Bod tání: 144…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075008
CAS číslo: 111061-44-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR