Tisk…

2'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2'-Amino-3-carboxybiphenyl hydrochloride, 3-(2-Aminophenyl)benzoic acid hydrochloride

OR5315-1G OR5315-250MG
APOSOR5315-1GEA 7940 CZK
APOSOR5315-1G APOSOR5315-250MG
2'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride
2'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride
Vzorec: C₁₃H₁₁NO₂·ClH
MW: 249,693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260476
CAS číslo: 1172351-47-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR