Tisk…

2-(4-Formylphenyl)acetonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR53129-1G OR53129-250MG
APOSOR53129-1GEA 16340 CZK
APOSOR53129-1G APOSOR53129-250MG
2-(4-Formylphenyl)acetonitrile 95%
2-(4-Formylphenyl)acetonitrile
Vzorec: C₉H₇NO
MW: 145,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756802
CAS číslo: 55211-74-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR