Tisk…

(Pyrimidin-2-yl)acetonitrile

Dodavatel: Apollo Scientific
Varování

OR53117-1G OR53117-5G
APOSOR53117-1GEA 8100 CZK
APOSOR53117-1G APOSOR53117-5G
(Pyrimidin-2-yl)acetonitrile
(Pyrimidin-2-yl)acetonitrile
Vzorec: C₆H₅N₃
MW: 119,124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759105
CAS číslo: 59566-45-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR