Tisk…

2,6-Dibromo-4-tert-butylphenol

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,6-Dibromo-4-(tert-butyl)phenol

OR53102-5G OR53102-25G
APOSOR53102-5GEA 2150 CZK
APOSOR53102-5G APOSOR53102-25G
2,6-Dibromo-4-tert-butylphenol
2,6-Dibromo-4-tert-butylphenol
Vzorec: C₁₀H₁₂Br₂O
MW: 308,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152389
CAS číslo: 98-22-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR