Tisk…

4-Bromo-6-methylpyridazin-3(2H)-one

Dodavatel: Apollo Scientific
Varování

OR53091-1G OR53091-250MG
APOSOR53091-1GEA 827 CZK
APOSOR53091-1G APOSOR53091-250MG
4-Bromo-6-methylpyridazin-3(2H)-one
4-Bromo-6-methylpyridazin-3(2H)-one
Vzorec: C₅H₅BrN₂O
MW: 189,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701342
CAS číslo: 954240-46-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR