Tisk…

(3R,4S,5R,6R)-3,4,5-tris(trimethylsilyl)oxy)-6-((trimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-2-one 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,3,4,6-Tetra-O-(trimethylsilyl)-D-glucono-delta-lactone, 2,3,4,6-tetra-O-(trimethylsilyl)-D-glucono-1,5-lactone, 2,3,4,6-tetrakis-O-trimethylsilyl-D-gluconolactone

OR53043-25G OR53043-100G OR53043-1KG
APOSOR53043-25GEA 1040 CZK
APOSOR53043-25G APOSOR53043-100G APOSOR53043-1KG
(3R,4S,5R,6R)-3,4,5-tris(trimethylsilyl)oxy)-6-((trimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-2-one 95%
(3R,4S,5R,6R)-3,4,5-tris(trimethylsilyl)oxy)-6-((trimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-2-one
Vzorec: C₁₈H₄₂O₆Si₄
MW: 466,864 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22665922
CAS číslo: 32384-65-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR