Di-tert-butyl chloromethyl phosphate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR53042-1G
OR53042-5G
OR53042-25G
APOSOR53042-1GEA
622
CZK
APOSOR53042-1G
APOSOR53042-5G
APOSOR53042-25G
Di-tert-butyl chloromethyl phosphate 95%
Di-tert-butyl chloromethyl phosphate
Vzorec:
C₉H₂₀ClO₄P MW: 258,679 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12165895 CAS číslo: 229625-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...