Tisk…

2-Ethynyl-5-methylpyrazine 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Ethynyl-5-methyl-1,4-diazine, (5-Methylpyrazin-2-yl)acetylene

OR53019-100MG OR53019-1G OR53019-250MG
APOSOR53019-100MGEA 2490 CZK
APOSOR53019-100MG APOSOR53019-1G APOSOR53019-250MG
2-Ethynyl-5-methylpyrazine 95+%
2-Ethynyl-5-methylpyrazine
Vzorec: C₇H₆N₂
MW: 118,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22199322
CAS číslo: 1207627-45-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR