2-Chloro-6-ethynylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR53011-1G
OR53011-250MG
APOSOR53011-1GEA
9920
CZK
APOSOR53011-1G
APOSOR53011-250MG
2-Chloro-6-ethynylpyridine
2-Chloro-6-ethynylpyridine
Vzorec:
C₇H₄ClN Storage Temperature: Ambient |
MDL Number:
MFCD13189719 CAS číslo: 914950-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...