5-Chloro-2-ethynylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5-Chloropyridin-2-yl)acetylene
OR53004-1G
OR53004-250MG
APOSOR53004-1GEA
6220
CZK
APOSOR53004-1G
APOSOR53004-250MG
5-Chloro-2-ethynylpyridine
5-Chloro-2-ethynylpyridine
Vzorec:
C₇H₄ClN MW: 137,566 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189715 CAS číslo: 1196153-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...